Please login to your online account to display your discounted pricing

Hanna™ Instruments Checker™ pH Tester and Replacement Part DFS Item

For use with virtually any electrode that shares same connector

$51.40 - $64.97

Products
Catalog Number Mfr. No. Price Quantity    

13-300-70

DFS Item
  Accessories
hanna instruments cda inc
HI98103
Each for $51.40

13-300-69

DFS Item
 
hanna instruments cda inc
HI1270
Each for $64.97
Description & Specifications

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.01 pH resolution at 0° to 50°C
  • Accuracy is ±0.2 pH
  • 1.5V
  • RH max 95%
  • With LCD and manual two point calibration
  • 3000 hours of battery life