Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, with phenol red


Quantity 500mL

Manufacturer: alfa aesar™  J67716AP

 View more versions of this product

Catalog No. AAJ67716AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
MDL Number MFCD00217418
Color Red
TSCA TSCA
Name Note without calcium, without magnesium, with phenol red
Physical Form Liquid
Quantity 500mL

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note without calcium, without magnesium, with phenol red
MDL Number MFCD00217418