Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), without calcium, without magnesium, with phenol red

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note without calcium, without magnesium, with phenol red
MDL Number MFCD00217418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67716AP

  SDS
alfa aesar™
J67716AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67716K2

  SDS
alfa aesar™
J67716K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
MDL Number MFCD00217418
Physical Form Liquid
TSCA TSCA
Name Note without calcium, without magnesium, with phenol red
Color Red
Quantity 500mL