Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), without calcium, with magnesium, with phenol red

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note without calcium, with magnesium, with phenol red
MDL Number MFCD00217418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67609AP

  SDS
alfa aesar™
J67609AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67609K2

  SDS
alfa aesar™
J67609K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Color Red
Name Note without calcium, with magnesium, with phenol red
MDL Number MFCD00217418
Physical Form Liquid
TSCA TSCA
Chemical Name or Material Hanks' Balanced Salt Solution (1X)