Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), with calcium, without magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note with calcium, without magnesium, without phenol red
MDL Number MFCD00217418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67807AP

  SDS
alfa aesar™
J67807AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67807K2

  SDS
alfa aesar™
J67807K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
MDL Number MFCD00217418
Physical Form Liquid
Quantity 500mL
Color Colorless
TSCA TSCA
Name Note with calcium, without magnesium, without phenol red