Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), with calcium, without magnesium, without phenol red


Quantity 1L

Manufacturer: alfa aesar™  J67807K2

 View more versions of this product

Catalog No. AAJ67807K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
MDL Number MFCD00217418
Physical Form Liquid
Color Colorless
Quantity 1L
TSCA TSCA
Name Note with calcium, without magnesium, without phenol red

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note with calcium, without magnesium, without phenol red
MDL Number MFCD00217418