Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (1X), with calcium, with magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
Name Note with calcium, with magnesium, without phenol red
MDL Number MFCD00217418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67763AP

  SDS
alfa aesar™
J67763AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67763K2

  SDS
alfa aesar™
J67763K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (1X)
MDL Number MFCD00217418
TSCA TSCA
Color Colorless
Name Note with calcium, with magnesium, without phenol red
Quantity 500mL
Physical Form Liquid