Please login to your online account to display your discounted pricing

Alfa Aesar™ Hanks' Balanced Salt Solution (10X), with calcium, with magnesium, without phenol red

Chemical Identifiers

Chemical Name or Material Hanks' Balanced Salt Solution (10X)
Name Note with calcium, with magnesium, without phenol red
MDL Number MFCD00217418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67771AP

  SDS
alfa aesar™
J67771AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67771K2

  SDS
alfa aesar™
J67771K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00217418
Chemical Name or Material Hanks' Balanced Salt Solution (10X)
Color Colorless
Quantity 500mL
TSCA TSCA
Name Note with calcium, with magnesium, without phenol red
Physical Form Liquid