Please login to your online account to display your discounted pricing

Hands-On Science Books


Explore scientific concepts through memorable activities based in the context of everyday life.

Manufacturer: fisher science education  0-37640

 View more versions of this product

Catalog No. S65932

  • / Each
 

Description & Specifications

Specifications

Includes One copy each of atmosphere and weather, rocks and minerals and our solar system
Product Type Atmosphere and Weather
Category Geography
Format Book
Grade 7 to 9