missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Scientific Hands-On Science Books
Explore scientific concepts through memorable activities based in the context of everyday life.
Supplier: Fisher Scientific 03935X
Description
- Easily integrated into any science course
- Correlate to the National Science Education Standards
- Grades 7 to 9
- Each book is 96 to 104 pages
ABCs of Chemistry
- Measure heat from a candle, make soap and build a simple battery
- Investigate factors affecting the speed of a reaction
- Learn to separate mixtures
- Plus 14 more activities
- The state we're in: Air is a gas!
- What is weather? Lightning!
- Convection: temperature differences and the motion of air, highs, lows and winds
- Plus 14 more activities
- Generate static electricity
- Explore magnetic fields, series circuits, conductors and insulators
- Create electricity from ice water
- Plus 11 more activities
- Making and adding vectors
- Velocity and acceleration
- Newton's 1st Law: Inertia
- Newton's 2nd Law: Fnet = ma
- Newton's 3rd Law: Action/reaction
- Pressure is not force
- Plus 11 more activities
- Get familiar with basic concepts, techniques and issues in biotechnology
- Making yogurt and kimchi
- Learn about gel electrophoresis and running a gel
- Investigate Building People and To Clone, or Not to Clone
- Plus 14 more acitivities
- What is the nature of light?
- How can we separate pigments?
- How do different materials affect the absorption of light?
- How light affects light-sensitive chemicals
- Plus 16 more activities
- Covers a variety of human anatomy and physiology topics such as: Seeing can be deceiving; Can eating ice cream be painful?; What parts of the body are involved in maintaining balance?; How much water does a popcorn kernel contain?; and milk, hot dogs and nutritional math
- The scale of the solar system: How big are the planets?
- How far apart are the planets?
- The shape of planetary orbits
- Plus 15 more activities
- Which way will the roots grow?
- Which color of light prevents chlorosis in geranium plants?
- How glucose content of a potato affects its use
- Plus 17 more activities
- Crystal structure of salt
- Mineral tests — hardness
- Rock types and mineral identification
- Weathering rates of rocks and minerals
- Plus 11 more activities
- Investigate: What is force? What is work? What is power?
- Mechanical advantages in a bicycle
- How a wedge and pulley work
- Plus 14 more activities
- How do ships float?
- When can water carry an electric current?
- How does water act like a lens?
- Investigate life in a pond
- Plus 16 more activities
Earth Science Series includes one copy each of Atmosphere & Weather, Rocks & Minerals and Our Solar System.
Life Science Series includes one copy each of Arthropods, Nutrition, Mind & Body and Plants.
Physical Science Series includes one copy each of Chemistry, Electricity & Magnetism, Force & Motion, Light & Color, Simple Machines and Water.
Specifications
George Watson | |
Book | |
Crystal structure of salt, mineral tests - hardness, mineral identification, rock types, weathering rates of rocks and minerals, 11 more activities, |
7 to 9 | |
Rocks and Minerals | |
Geography |