Foss™ Hammertec™ Sample Mill with Feeder
Designed for falling number analysis, wet gluten analysis, Kjeldahl and NIR analysis. Foss North America Hammertec Sample Mill is suitable for dry, flowable whole cereal grain samples.
Supplier:  Foss™ 80350002
Catalog No. 12-053-138
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: Hammertec, complete with Auto-feeder, sample outlet adaptor, sample funnel with adaptor, flat belt, sample bag, sample brusher, 0.8mm sample filter, package box, owner' guide, spare parts manual, manual box
Description
Description
  • Ideal for sample preparation where requirements for fineness and uniformity of particle size are high
  • Small and light making it easier to move
  • 60% smaller than equivalent mills taking up less room on your bench
  • Low noise eliminates need to use ear protection
Specifications
Specifications
Sample Mill
100/120 V, 50/60 Hz, 1300 W
300 g in less than 60 sec. at 13% moisture level, depending on sample type and moisture content
Hammertec, complete with Auto-feeder, sample outlet adaptor, sample funnel with adaptor, flat belt, sample bag, sample brusher, 0.8mm sample filter, package box, owner' guide, spare parts manual, manual box
Up to 10 mm (up to 40 mm with large inlet)
16,800 rpm
40.6 kg
55.5 x 24 x 46 cm
Falling number analysis, wet gluten analysis, Kjeldahl and NIR analysis
50/60 Hz
Hammertec
Dry, flowable whole cereal grain samples
100/120 V