Hamilton™ Syringes for Agilent HP7673 and HP7683 Autosamplers
For use with Agilent Technologies HP7673, HP7683, HP7670, HP7671, and HP7672 autosamplers
$85.11 - $85.11
Specifications
Type Gastight™ Syringe
Volume (Metric) 5μL
For Use With (Equipment) Agilent Technologies HP7673, HP7683, HP7670, HP7671, and HP7672 autosamplers
Length (English) Needle 1.71 in.
Length (Metric) Needle 43.4mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Connection Type Model Quantity Needle Gauge Price Quantity  
Catalog Number Mfr. No. Connection Type Model Quantity Needle Gauge Price Quantity  
14-813-260 Hamilton™
HAM80076
Cemented 75ASN Each 23s-26s ga Each for $85.11
Specifications
Description
  • Available with three needle gauges: 23s/26s, 23s, and 26s, and two needle point styles: Style 2 and Style HP
  • Style 2 is a beveled, noncoring needle point for septum penetration
  • Style HP is a special conical style used exclusively on syringes for the Agilent Technologies autosamplers
  • Needles are 1.71 in. (4.3cm) long
Description & Specifications
Specifications
Gastight™ Syringe
Agilent Technologies HP7673, HP7683, HP7670, HP7671, and HP7672 autosamplers
43.4mm
5μL
1.71 in.
Cone