missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hamilton™ Syringe from Restek
High precision and accuracy down to 0.1μL.
Supplier: Chromatographic RK24555
Description
- Sample is contained in the needle with full plunger displacement, ensuring no dead volume
- Seal between the plunger and the needle can be tightened for high-pressure injection
- Replaceable needle/plunger kits and barrels
- Needle Length: 70mm (2.75 in.)
Specifications
PTFE (Tip) | |
For high pressure injection | |
Hamilton Micro-Volume, Positive Displacement Syringes | |
2 | |
5.0 μL | |
Knurled Hub |
24 G | |
2.75 in. | |
7105 | |
Ea. | |
2 |