missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamilton™ Small Hub Removable Needles (2.5 - 100μL Syringes)

Can be used with removable (RN)-style syringes that are 100μL and smaller.

Manufacturer:  Hamilton™ HAM776204

 View more versions of this product

Catalog No. 14-824-711



Description

Description

  • Standard needle length is 51mm (2 in.)
  • Custom gauges, lengths and point styles are available
Specifications

Specifications

Autoclavable
0.005 in.
0.13 mm
Ideal when there is a risk of the needle clogging
51 mm
Blunt
31 G
0.01 in.
0.26 mm
2 in.
Stainless Steel
Replacement Syringe Needle
Documents