missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hamilton™ PRP™-X200 Cation Exchange Columns
Analyze inorganic and organic cations using conductivity or UV detection. Separate mono or divalent cations depending on mobile phase conditions from 20 ppb to 200 ppm.
Supplier: Hamilton™ HAM79441
Description
Can be used with any existing HPLC or ion chromatograph to easily determine cations in air, water and soil. PRP-X200 columns provide a number of benefits
Better than Wet Chemical Methods
- Ion chromatography will greatly reduce sample pretreatment and improve the accuracy and precision of results.
- Separate ions at concentrations from 20ppb to 20ppm without suppression or elaborate preconcentration techniques
- Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), a regeneration protocol will usually return your chromatography to its original state.
- Quickly and easily separate five monovalent cations in five minutes or four divalent cations in four minutes
- Highly inert poly(styrene-divinylbenzene) packing allows the use of organic modifier. By varying the concentration of methanol the resolution between cations can be altered.
- One particle size: 10°m
- Four column internal diameters: 2.1 to 10.0mm
- Two column materials: 316 stainless steel and PEEK
- Analytical and semiprep/preparative guard columns
Specifications
Analytical Column | |
2 mL/min | |
150 mm | |
pH 1 to 13, 0 to 100% aqueous, organic modifier | |
1 to 13 | |
5°C to 60°C | |
PRP-X200 | |
Ion Exchange | |
Stainless Steel |
4.1 mm | |
Analyzing Inorganic and Organic Cations Using Conductivity or UV Detection | |
5000 psi (345 bar) | |
10 μm | |
100 Å | |
L22 | |
PSDVB/Sulfonic Acid | |
Ion Exchange | |
Column |