Please login to your online account to display your discounted pricing
Hamilton™ PRP™-X100 Guard Cartridge Kit
Stainless steel and PEEK guard columns help protect your investment
$337.11 - $471.94
Specifications
For Use With Virtually all Solvents except high concentrations of Sodium Chloride
Max. Pressure 5000psi
Particle Size 10μm
View More Specs
Products
Catalog Number Mfr. No. Column Capacity Material Length (Metric) Price Quantity    
14-816-120
hamilton™
HAM79383
57μL Each for $471.94
14-816-22
hamilton™
HAM79448
80μL Each for $337.11
Description & Specifications
Specifications
For Use With Virtually all Solvents except high concentrations of Sodium Chloride
Max. Pressure 5000psi
Particle Size 10μm
  • Guard columns remove particulate contaminants and highly adsorptive compounds from samples
  • Modular and cartridge replacement is easy and tool free
  • Holders are available in stainless steel and PEEK materials, both designs optimized to reduce dead volume and prevent extra column band broadening
  • Holders for the cartridges are reusable indefinitely
Kit contains 1 holder and 2 cartridges
Recommended for use with virtually all solvents except high concentrations of Sodium Chloride.