Hamilton™ PRP-C18 Reversed-Phase HPLC Column Accessories - Bulk Resin
Bulk resin for individuals who prefer to pack their own HPLC columns
Manufacturer:  Hamilton™ HAM79793
 View more versions of this product
Catalog No. 13-622-121
Description
Description
Designed to provide high-efficiency, reversed-phase separations over an extended column life in nearly any mobile phase or pH. The rigid stationary phase has excellent mechanical and thermal stability, does not experience shrinking or swelling, and is completely inert to most conditions commonly encountered in reversed-phase chromatography. Hamilton PRP-C18 bulk resin is for general purpose HPLC and is especially well-suited for specialized applications in the clinical, pharmaceutical, environmental, food, forensics, and life sciences industries.
Benefits & Features
  • pH stable from 1 to 14
  • Temperature stability above 100°C
  • Unique bonding of C18 to a polymer support
  • LC/MS applications, synthesized DNA, and small molecules
  • Excellent durability (stable to any concentration of water or organic solvent)
Specifications
Specifications
PRP-C18
12 to 20μm
1g
>100°C
1 to 14
Documents