MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Cone Tip, ASRN (Autosampler Removable Needle), Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Manufacturer:  MilliporeSigma™ Supelco™ 24582
Catalog No. 111011098
Description
Description
Specifications
Specifications
1.69 in.
6/Pk.
Cone Tip, ASRN (Autosampler Removable Needle)
Hamilton RN Syringes
Stainless Steel
5 to 10 µL
43 mm
SDS
Documents
Product Certifications
Promotions
Promotions
Hamilton is a registered trademark of Hamilton Co.