Hamilton™ Tungsten Cleaning Wires
For mechanical cleaning to remove needle obstructions.
Manufacturer:  Hamilton™ HAM18304
 View more versions of this product
Catalog No. 14-819-79
Description
Description
Tungsten wires, best used with Hamilton Company Needle Cleaning Concentrate (14-819-85, 14-819-86).
Order Info
Ten wires per pack.
Specifications
Specifications
22, 23G, and larger needle, syringe and needle cleaning kit
17.78cm
Cleaning Wire
7 in.
Tungsten
Documents