Hamilton™ Tungsten Cleaning Wires
For mechanical cleaning to remove needle obstructions.
Manufacturer:  Hamilton™ HAM18303
 View more versions of this product
Catalog No. 14-819-78
Description
Description
Tungsten wires, best used with Hamilton Company Needle Cleaning Concentrate (14-819-85, 14-819-86).
Order Info
Ten wires per pack.
Specifications
Specifications
24 to 26G needle, syringe and needle cleaning kit
7 in.
Tungsten
Cleaning Wire
17.78 cm
Documents