Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Manufacturer:  Hamilton™ HAM6149101
 View more versions of this product
Catalog No. 14-816-267
Description
Description
  • For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
Specifications
Microlab™ 600 diluter/dispenser system
FEP
3.75μL/mm
1/4-28 Tubing threads
Gauge 12, Continuous Fill Tubing
Documents