Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
Manufacturer:  Hamilton™ HAM5900055
 View more versions of this product
Catalog No. 14-816-279
Description
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Specifications
Microlab™ 600 dispensers
Bubble free prime
25mL
Documents