Hamilton™ Microlab 300 Pipetting System Accessories - Syringes
Choose from two syringes designed for use specifically with the Hamilton Microlab 300 Pipetting System
Manufacturer:  Hamilton™ HAM6608815
 View more versions of this product
Catalog No. 14-815-917
Description
Description
Model 1705.5
  • Bubble Free Prime
  • Expels air from the syringe and valve thanks to an innovative plunger tip
  • 1.25mL
  • Model 1002.5
    • Gastight
    • Ideal for dispensing both liquids and gases
    • Has a precision-machined PTFE plunger tip which creates a leak-free seal
    • 25μL
Specifications
Specifications
Microlab™ 300 pipetteting system
Plunger tip
1705.5 BFP
25 μL
Documents