Hamilton™ Microlab™ 1700 Series Gastight™ Syringes
For dispensing both liquids and gases
Supplier: Hamilton™ HAM80062
Description
- Equipped with precision-machined polymer plunger tip, often PTFE, which creates leak-free seal
- Tight fit feature ensures tip wipes interior of syringe barrel free of sample
- Reduces the chance deposit will occur in samples with dissolved or suspended molecules and result in damaged plunger
- ChemSeal termination
Specifications
26s G | |
1701 | |
10 μL |