Hamilton™ Metal Hub Custom Needles (Luer Lock)
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Custom needle lengths and point styles are available.
Manufacturer:  Hamilton™ HAM774804
Catalog No. 22-288678
Description
Description
Metal Hub Compatible Syringe Terminations
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life
  • Needle Point Styles
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 33-10
    • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 33-10
    • Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 33-10. Special needle styles such as 30° or 45° are available upon request
    Six needles per pack.
    Order Info
    Needle Length – Must be specified on a per order basis. 0.375-12 in. lengths are available
    Needle Point Style – Must be specified on a per order basis
    Specifications
    Specifications
    Autoclavable
    0.03 in.
    0.84 mm
    Replacement Needle
    18 G
    0.05 in.
    1.27 mm
    TLL syringes and LT or TLL connectors