Hamilton™ Luer Lock Adapter for Disposable Tips
$104.74 - $104.74
Specifications
For Use With (Equipment) PB600-1 repeating dispensers with disposable tip, ideal eliminating the possibility cross contamination
Material CTFE
Type Luer-tip Adapter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
13-684-78F Hamilton™
HAM31330
100μL Each for $104.74
13-684-78G Hamilton™
HAM31331
1000μL Each for $104.74
Specifications
Description
  • Two luer tip adapters available: 100μL and 1mL
Description & Specifications
Specifications
PB600-1 repeating dispensers with disposable tip, ideal eliminating the possibility cross contamination
CTFE
Luer-tip Adapter