Hamilton™ Gastight™ Spark Holland HPLC Autosampler Syringes
Feature rugged polytetrafluoroethylene (PTFE) plunger tip and are economically designed to offer long-lasting, top-level quality
Manufacturer:  Hamilton™ HAM5465901
 View more versions of this product
Catalog No. 13-621-731
Description
Description
Strictly machined tolerances for reproducible results from injection-to-injection, and their glass barrel surface finish provides superior syringe life.
The Hamilton Spark Holland syringe line works with the following autosamplers: Spark Holland Midas Triathlon, Endurance, Alias, Integrity, Optimas; Varian ProStar 410, ProStar 420, ProStar 430; Dionex Model 542, 540, 540MT; Perkin Elmer Series 225, FLEXAR; Beckman Coulter 508; Grace/Alltech 580; Antec AS 100, AS 110; Knauer Smartline 3950; PLATINblue AS-1; KTA A 900, A 905.
Specifications
Specifications
0.091 in.
0.3 in.
0.77cm
Spark Holland midas triathlon, endurance, alias, integrity, optimas, varian prostar 410, prostar 420, prostar 430, dionex model 542, 540, 540MT, PerkinElmer™ series 225, FLEXAR, Beckman coulter 508, grace/alltech 580, antec AS 100, AS 11
No Needle Included
239°F
50°F
No Needle Included
N/A
Gastight™ Autosampler Syringe
No needle included
0.23cm
250μL
No Needle Included
68.9 bar
115°C
1725C
10°C
Each
Documents