Hamilton™ Constant Rate-700 HPLC Syringes & Replacement Parts
Syringes are designed to delive reasy, reproducible injection of liquid samples into an injector on HPLC instrumentation. The CR-700 standardizes three major variables: sample volume, rate of injection, and depth of needle penetration. Even when operated by different technicians, the CR-700 provides reproducibility of +1% of the total syringe volume in routine procedures.
Manufacturer:  Hamilton™ HAM15936
 View more versions of this product
Catalog No. 14-823-5
Description
Description
Volumes are selected with micrometer precisions, the plunger is spring-driven and injects the samples at a constant rate. Three models of the CR-700 syringes are available with volumes of 20, 50 or 200μL.
Needle Point Styles
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauge: 22
Specifications
Specifications
22 G
2 in.
CR-700
Each
HPLC injection valves
51mm
Blunt
Replacement Needle
Documents