missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamilton™ 700 Microliter™ Syringes: RN Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.

Manufacturer:  Hamilton™ HAM87930

 View more versions of this product

Catalog No. 14-824-53B



Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications

Specifications

Removable
0.014 in.
0.13 mm
0.26 in.
6.6 mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
5.1 cm
PTFE
239°F
50°F
2
Each
5 µL
26s G
0.005 in.
0.019 in.
0.343 mm
0.483 mm
2 in.
51 mm
Stainless Steel
2000 psi
115°C
75
Sharp, Bevel, Curved, Non-coring
Microliter Syringe
Documents