missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamilton™ 700 Microliter™ Syringes: RN Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.

Manufacturer:  Hamilton™ HAM80336

 View more versions of this product

Catalog No. 14-824-1



Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications

Specifications

0.005 in.
0.019 in.
0.13mm
0.26 in.
6.6mm
10μL
2 in.
2 in.
Stainless Steel
2000psi
115°C
701
Sharp, Bevel, Curved, Non-coring
Microliter™ Syringe
Removable
26s G
0.0187 in.
0.485mm
0.47mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
51mm
5.1cm
PTFE
239°F
50°F
2
6/Pk.
Documents