missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamilton™ 700 Microliter™ Syringes: LT Termination

The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 500 µL. A variety of different terminations and needle options are available in this series.

Manufacturer:  Hamilton™ HAM80401

 View more versions of this product

Catalog No. 14-813-18



Description

Description

  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications

Specifications

Yes
No Needle Included
0.729mm
25μL
No Needle Included
Stainless Steel
2000psi
115°C
702
N/A
Luer-tip
0.029 in.
0.305 in.
7.75mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
No Needle Included
PTFE
239°F
50°F
No Needle Included
Each
Microliter™ Syringe
Documents