Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: DAD Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
$202.59 - $224.37
Specifications
Type Gastight™ Syringe
For Use With (Equipment) For Microlab™ 1000 diluters/dispensers and features a side port connector
Needle Gauge No needle included
Material (Barrel) Borosilicate Glass
Model 1705
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Autoclavable Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Autoclavable Volume (Metric) Price Quantity  
13-684-79EE Hamilton™
HAM201600
Yes 250μL Each for $216.60
13-684-79AA Hamilton™
HAM201500
Yes 50μL Each for $202.59
13-684-79JJ Hamilton™
HAM201700
No 1.0mL Each for $224.37
Specifications
Description
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • Diluter AccuDil (DAD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers and features a side port connector.
Description & Specifications
Specifications
Gastight™ Syringe
No needle included
1705
Diluter AccuDil™
For Microlab™ 1000 diluters/dispensers and features a side port connector
Borosilicate Glass
Each