Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: DAD Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Manufacturer:  Hamilton™ HAM201500
 View more versions of this product
Catalog No. 13-684-79AA
Description
Description
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • Diluter AccuDil (DAD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers and features a side port connector.
Specifications
Specifications
Yes
50μL
No Needle Included
Borosilicate Glass
1000psi
50°C
1705
N/A
Diluter AccuDil™
No needle included
For Microlab™ 1000 diluters/dispensers and features a side port connector
No Needle Included
PTFE
122°F
68°F
No Needle Included
Each
Gastight™ Syringe
Documents