Hamilton™ 1700/1000 Series Gastight™ Instrument Syringes: AD Termination
Syringes are designed to fit into, or with, various different instruments such as the MICROLAB™ 500 and MICROLAB 1000
Manufacturer:  Hamilton™ HAM201100
 View more versions of this product
Catalog No. 13-684-79G
Description
Description
Have a PTFE plunger tip that creates a tight connection between the syringe barrel and plunger allowing for liquid or gas sampling.
  • AccuDil™ (AD) - This termination is designed for MICROLAB 1000 Diluters/Dispensers
Specifications
Specifications
No needle included
For Microlab™ 1000 diluters/dispensers
No Needle Included
PTFE
122°F
68°F
No Needle Included
Each
Gastight™ Syringe
250μL
No Needle Included
Borosilicate Glass
500psi
50°C
1725
N/A
AccuDil™
Documents