Please login to your online account to display your discounted pricing

Thermo Scientific™ Halt™ Protease Inhibitor Cocktails


Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.

Manufacturer: thermo scientific™  87785

 View more versions of this product

Catalog No. PI87785

  • / Each
 

Description & Specifications

Specifications

Quantity 1mL
Sufficient For 100mL of sample
Composition AEBSF-HCl, Aprotinin, Bestatin, E64, Leupeptin, Pepstatin A
Formulation Inhibitors stabilized in DMSO
Description Halt Protease Inhibitor Cocktail, EDTA-Free
Format Liquid

Thermo Scientific™ Halt Protease Inhibitor Cocktails are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction procedures.

Halt Protease Inhibitor Cocktails are 100X solutions containing optimized concentrations of six broad-spectrum protease inhibitors stabilized in high-quality dimethylsulfoxide (DMSO). The cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The identity and concentration of each component is fully disclosed, leaving no uncertainties about variables and compatibility with experiments. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively. The stable, concentrated liquid cocktails are easy to use, and the several different package sizes make Halt Protease Inhibitor Cocktails exceptionally convenient for either small-scale or large-scale uses.

Highlights:

  • Better than tablets – the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need

  • No proprietary ingredients – fully disclosed formulation contains AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A in DMSO

  • Multiple package sizes – choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination

  • Two popular formulations – regular formulation includes a separate vial of EDTA solution for optional metalloprotease inhibition; EDTA-free formulation ensures compatibility with isoelectric focusing

  • Lysis buffer compatible – use with Thermo Scientific Pierce™ Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent

Includes:

Protease inhibitor cocktails with EDTA include separate 0.5M EDTA solution.