Please login to your online account to display your discounted pricing

Haloperidol, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0215369694

 View more versions of this product

Catalog No. ICN15369694

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical State Solid
Quantity 5g
Chemical Name or Material Haloperidol
CAS 52-86-8
Physical Form White powder
Molecular Formula C21H23CIFNO2
Formula Weight 375.88g/mol (anhydrous)
MDL Number MFCD00051423
Synonym 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone
Recommended Storage Store at Room Temperature (15° to 30°C).
EINECS Number 200-155-6

Specifications

Chemical Name or Material Haloperidol
CAS 52-86-8
Physical Form White powder
Molecular Formula C21H23CIFNO2
Formula Weight 375.88g/mol (anhydrous)
MDL Number MFCD00051423
Synonym 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone

Antagonist

Accessories

Store at Room Temperature (15° to 30°C).