Please login to your online account to display your discounted pricing

Haloperidol, MP Biomedicals™

$123.67 - $123.67

Chemical Identifiers

Chemical Name or Material Haloperidol
CAS 52-86-8
Physical Form White powder
Molecular Formula C21H23CIFNO2
Formula Weight 375.88g/mol (anhydrous)
MDL Number MFCD00051423
Synonym 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15369694

 
mp biomedicals
0215369694
5g Each for $123.67
Description & Specifications

Specifications

Physical State Solid
Quantity 5g
Chemical Name or Material Haloperidol
CAS 52-86-8
Physical Form White powder
Molecular Formula C21H23CIFNO2
Formula Weight 375.88g/mol (anhydrous)
MDL Number MFCD00051423
Synonym 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone
Recommended Storage Store at Room Temperature (15° to 30°C).
EINECS Number 200-155-6

Antagonist

Safety and Handling

Store at Room Temperature (15° to 30°C).