Please login to your online account to display your discounted pricing

Alfa Aesar™ Haloenol Lactone Suicide Substrate, ≥98%

For cellular and molecular biology applications

Specifications

Storage Requirements -30° to -10°C
For Use With (Application) Irreversibly inhibits calcium-independent phospholipase A-2
Color White
Molecular Weight 317.18
Format Lyophilized
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Substrate Quantity Price Quantity    

AAJ63728LB0

 
alfa aesar™
J63728LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Storage Requirements -30° to -10°C
For Use With (Application) Irreversibly inhibits calcium-independent phospholipase A-2
Color White
Molecular Weight 317.18
Format Lyophilized