missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HAI-1 Mouse anti-Human, Clone: 9B10, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $158.05

Specifications

Antigen HAI-1
Clone 9B10
Host Species Murine
Gene Alias HGF activator inhibitor-1, hepatocyte growth factor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013925 Life Technologies
LS14996080
25μg Each for $100.05
5013926 Life Technologies
LS14996082
100μg Each for $158.05
Specifications

Description

The monoclonal antibody 9B10 recognizes HAI-1 also known as hepatocyte growth factor activation inhibitor. HAI-1 is a single transmembrane protein of approximately 53kDa with an extracellular Kunitz-type serine protease inhibitor domain. HAI-1 and HAI-2 are produced in membrane-associated forms, which are secreted as active, proteolytically truncated proteins. HAI can bind to and inhibit HGFA and matripase, which are responsible for converting HGF to an active form. Human HAI-1 transcript is expressed in various human tissues, such as adult placenta, kidney, pancreas, prostate, small intestine, fetal kidney and fetal lung. HAI-1 is a marker for prostate and breast cancers.
Description & Specifications

Specifications

HAI-1
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
9B10
HGF activator inhibitor-1, hepatocyte growth factor
Flow Cytometry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL