missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium wire, 2.0mm (0.08 in.) dia., 99.7% (metals basis excluding Zr), Zr nominal 3%

Manufacturer:  Alfa Aesar™ 13305BY

 View more versions of this product

Catalog No. AA13305BY



Specifications

Specifications

Hafnium wire
100cm
7440-58-6
Hf
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
[Hf]
Odorless
23986
Wire
≈41.1g/m
2.0mm (0.08 in.) dia.
(metals basis excluding Zr), Zr nominal 3%
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
178.49
CHEBI:33343
99.7%

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents