missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03922306

 View more versions of this product

Catalog No. AA3922306



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hafnium telluride
Powder
(metals basis)
MFCD00049947
BCMXGUYIVFZMCT-UHFFFAOYSA-N
bis(tellanylidene)hafnium
90471877
99.5%
39082-23-0
5 g
HfTe2
hafnium telluride, ditellanylidenehafnium
[Te]=[Hf]=[Te]
433.69
433.69

Chemical Identifiers

39082-23-0
433.69
BCMXGUYIVFZMCT-UHFFFAOYSA-N
90471877
[Te]=[Hf]=[Te]
HfTe2
MFCD00049947
hafnium telluride, ditellanylidenehafnium
bis(tellanylidene)hafnium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 254-280-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only