Learn More
Hafnium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLHF12M
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Hf | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Hafnium Standard | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Hf | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
hafnium | |
23986 | |
2% HCl |
7440-58-6 | |
30 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011032 | |
[Hf] | |
178.49 | |
CHEBI:33343 |
Chemical Identifiers
7440-58-6 | |
178.49 | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
CHEBI:33343 | |
[Hf] |
Hf | |
MFCD00011032 | |
23986 | |
hafnium |
Safety and Handling
Warning: Causes skin irritation, serious eye irritation Wear protective gloves/eye/face protection. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If skin/eye irritation occurs, get medical advice/attention.
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only