Please login to your online account to display your discounted pricing

Hafnium sponge, 99.6% (metals basis excluding Zr), Alfa Aesar™

$74.28 - $74.28

Chemical Identifiers

Chemical Name or Material Hafnium
CAS 7440-58-6
Physical Form Sponge, 0.8-20mm (0.03-0.79 in.)
Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
MDL Number MFCD00011032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA0067409

  SDS
alfa aesar
0067409
10g Each for $74.28

AA0067418

  SDS
alfa aesar
0067418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA0067430

  SDS
alfa aesar
0067430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA00674A1

  SDS
alfa aesar
00674A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Color Silver-gray
DOT Information DOT Class: 4.2, Packaging Group: II
EINECS Number 231-166-4
Physical Form Sponge, 0.8-20mm (0.03-0.79 in.)
MDL Number MFCD00011032
CAS 7440-58-6
Odor Odorless
Appearance Silver gray sponge
Chemical Name or Material Hafnium
Assay Percent Range 99.6%, Zr nominal 3% (Metals basis excluding Zr)
Health Hazard 1 H251: Self-heating: may catch fire.
Safety and Handling

H251: Self-heating: may catch fire.