missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium silicide

Manufacturer:  Alfa Aesar™ 1256404

 View more versions of this product

Catalog No. AA1256404



Specifications

Specifications

Hafnium silicide
HfSi2
hafnium silicide, hafnium silicide
[Si].[Si].[Hf]
234.66
234.66
Odorless
2g
12401-56-8
MFCD00167026
TWRSDLOICOIGRH-UHFFFAOYSA-N
hafnium;silicon
11230071
Powder
1680°C

Chemical Identifiers

12401-56-8
234.66
TWRSDLOICOIGRH-UHFFFAOYSA-N
11230071
[Si].[Si].[Hf]
HfSi2
MFCD00167026
hafnium silicide, hafnium silicide
hafnium;silicon
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents