missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium rod, 6.35mm (0.25 in.) dia., 99.9% (metals basis excluding Zr), Zr nominal 3%

Manufacturer:  Alfa Aesar™ 44489DJ

 View more versions of this product

Catalog No. AA44489DJ



Specifications

Specifications

Hafnium rod
6.35mm (0.25 in.) dia.
(metals basis excluding Zr), Zr nominal 3%
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
178.49
CHEBI:33343
99.9%
50mm
7440-58-6
Hf
and compounds
[Hf]
Odorless
23986
Rod

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
and compounds
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

EINECSNumber : 231-166-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents