missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium powder, -325 mesh, 99.6% (metals basis excluding Zr), Zr nominal 2-3.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01020114

 View more versions of this product

Catalog No. AA1020114



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-58-6
Hf
UN2545
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
[Hf]
178.49
CHEBI:33343
−325 Mesh
Powder
Hafnium powder
(metals basis excluding Zr), Zr nominal 2 to 3.5%
MFCD00011032
25 g
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
23986
99.6%
Odorless

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

GHS H Statement
H251
Self-heating: may catch fire.

P210-P222-P280-P335+P334-P370+P378q

H250

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY

EINECSNumber : 231-166-4

RTECSNumber : MG4600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only