missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium powder, -325 mesh, 99.6% (metals basis excluding Zr), Zr nominal 2-3.5%

Manufacturer:  Alfa Aesar™ 1020122

 View more versions of this product

Catalog No. AA1020122



Specifications

Specifications

Hafnium powder
7440-58-6
Hf
UN2545
and compounds
[Hf]
Odorless
23986
Powder
Gray
100g
(metals basis excluding Zr), Zr nominal 2 to 3.5%
MFCD00011032
−325 Mesh
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
178.49
CHEBI:33343
99.6%

Chemical Identifiers

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
23986
hafnium
Hf
MFCD00011032
and compounds
CHEBI:33343
[Hf]
Safety and Handling

Safety and Handling

GHS H Statement
H251
Self-heating: may catch fire.

GHS P Statement
P280-P235+P410-P420-P407
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Store away from other materials.
Maintain air gap between stacks/pallets.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY

RTECSNumber : MG4600000

TSCA : TSCA

Documents