Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium powder, -100 mesh, 99.8% (metals basis excluding Zr), Zr nominal <4.5%

Chemical Identifiers

Chemical Name or Material Hafnium
CAS 7440-58-6
MDL Number MFCD00011032
UN Number UN2545
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA7774922

  SDS
alfa aesar™
7774922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

DOT Information Transport Hazard Class: 4.2; Packaging Group: II; Proper Shipping Name: HAFNIUM POWDER, DRY
Mesh Size −100 mesh
TSCA TSCA
Assay 99.8%
Health Hazard 3 GHS P Statement
P280-P235+P410-P420-P407
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Store away from other materials.
Maintain air gap between stacks/pallets.
Odor Odorless
Health Hazard 1 Danger
MDL Number MFCD00011032
UN Number UN2545
Assay Percent Notes (metals basis excluding Zr), Zr nominal <4.5%
CAS 7440-58-6
Health Hazard 2 GHS H Statement
H251
Self-heating
may catch fire.
Quantity 100g
RTECS Number MG4600000
Chemical Name or Material Hafnium
Safety and Handling

Danger

GHS P Statement
P280-P235+P410-P420-P407
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Store away from other materials.
Maintain air gap between stacks/pallets.

GHS H Statement
H251
Self-heating
may catch fire.