Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) oxide, tech.

Chemical Identifiers

Chemical Name or Material Hafnium(IV) oxide
CAS 12055-23-1
Purity Grade Technical Grade
Formula Weight 210.49
MDL Number MFCD00003565
Merck Index 14,4588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4027022

 
alfa aesar™
4027022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4027036

 
alfa aesar™
4027036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.49
Chemical Name or Material Hafnium(IV) oxide
TSCA TSCA
Mesh Size −325 Mesh
Odor Odorless
Purity Grade Technical Grade
Physical Form Powder
MDL Number MFCD00003565
CAS 12055-23-1
Merck Index 14,4588
Melting Point 2774°C
Quantity 100g
Density 9.68g/mL