Please login to your online account to display your discounted pricing

Alfa Aesar™ Hafnium(IV) oxide sputtering target, 101.4mm (4.0 in.) dia x 6.35mm (0.25 in.) thick, 99.95% (metals basis excluding Zr)

Chemical Identifiers

Chemical Name or Material Hafnium (IV) oxide sputtering target
Name Note 101.4mm (4.0 in.) dia. x 6.35mm (0.25 in.) thick
Molecular Formula HfO2
MDL Number MFCD00003565
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44875KS

  SDS
alfa aesar™
44875KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hafnium (IV) oxide sputtering target
TSCA TSCA
Odor Odorless
Name Note 101.4mm (4.0 in.) dia. x 6.35mm (0.25 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis excluding Zr)
Molecular Formula HfO2
MDL Number MFCD00003565
Quantity 1 each